MilliporeSigma
  • Apoptotic death of hematopoietic tumor cells through potentiated and sustained adhesion to fibronectin via VLA-4.
Apoptotic death of hematopoietic tumor cells through potentiated and sustained adhesion to fibronectin via VLA-4.
The Journal of biological chemistry (2009-12-17)
Yohei Saito, Toshiyuki Owaki, Takuya Matsunaga, Mizue Saze, Shogo Miura, Mao Maeda, Mayu Eguchi, Rika Tanaka, Junichi Taira, Hiroaki Kodama, Sumio Goto, Yoshiroh Niitsu, Hiroshi Terada, Fumio Fukai
ABSTRACT
It has been postulated that inactivated beta1-integrins are involved in the disordered growth of hematopoietic tumor cells. We recently found that TNIIIA2, a peptide derived from tenascin-C, strongly activates beta1-integrins through binding with syndecan-4. We show here that Ramos Burkitt's lymphoma cells can survive and grow in suspension but undergo apoptosis when kept adhering to fibronectin by stimulation with TNIIIA2. Other integrin activators, Mg(2+) and TS2/16 (an integrin-activating antibody), were also capable of inducing apoptosis. The inactivation of ERK1/2 and Akt and the subsequent activation of Bad were involved in the apoptosis. The results using other hematopoietic tumor cell lines expressing different levels of fibronectin receptors (VLA-4 and VLA-5) showed that potentiated and sustained adhesion to fibronectin via VLA-4 causally induces apoptosis also in various types of hematopoietic tumor cells in addition to Ramos cells. Because TNIIIA2 requires syndecan-4 as a membrane receptor for activation of beta1-integrins, it induced apoptosis preferentially in hematopoietic tumor cells, which expressed both VLA-4 and syndecan-4 as membrane receptors mediating the effects of fibronectin and TNIIIA2, respectively. Therefore, normal peripheral blood cells, such as neutrophils, monocytes, and lymphocytes, which poorly expressed syndecan-4, were almost insusceptible to TNIIIA2-induced apoptosis. The TNIIIA2-related matricryptic site of TN-C could contribute, once exposed, to preventing prolonged survival of hematopoietic malignant progenitors through potentiated and sustained activation of VLA-4.
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse