MilliporeSigma
  • Protective role of ALDH2 against acetaldehyde-derived DNA damage in oesophageal squamous epithelium.
Protective role of ALDH2 against acetaldehyde-derived DNA damage in oesophageal squamous epithelium.
Scientific reports (2015-09-17)
Yusuke Amanuma, Shinya Ohashi, Yoshiro Itatani, Mihoko Tsurumaki, Shun Matsuda, Osamu Kikuchi, Yukie Nakai, Shin'ichi Miyamoto, Tsunehiro Oyama, Toshihiro Kawamoto, Kelly A Whelan, Hiroshi Nakagawa, Tsutomu Chiba, Tomonari Matsuda, Manabu Muto
ABSTRACT
Acetaldehyde is an ethanol-derived definite carcinogen that causes oesophageal squamous cell carcinoma (ESCC). Aldehyde dehydrogenase 2 (ALDH2) is a key enzyme that eliminates acetaldehyde, and impairment of ALDH2 increases the risk of ESCC. ALDH2 is produced in various tissues including the liver, heart, and kidney, but the generation and functional roles of ALDH2 in the oesophagus remain elusive. Here, we report that ethanol drinking increased ALDH2 production in the oesophagus of wild-type mice. Notably, levels of acetaldehyde-derived DNA damage represented by N(2)-ethylidene-2'-deoxyguanosine were higher in the oesophagus of Aldh2-knockout mice than in wild-type mice upon ethanol consumption. In vitro experiments revealed that acetaldehyde induced ALDH2 production in both mouse and human oesophageal keratinocytes. Furthermore, the N(2)-ethylidene-2'-deoxyguanosine levels increased in both Aldh2-knockout mouse keratinocytes and ALDH2-knockdown human keratinocytes treated with acetaldehyde. Conversely, forced production of ALDH2 sharply diminished the N(2)-ethylidene-2'-deoxyguanosine levels. Our findings provide new insight into the preventive role of oesophageal ALDH2 against acetaldehyde-derived DNA damage.