MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • IL-17RE is the functional receptor for IL-17C and mediates mucosal immunity to infection with intestinal pathogens.
IL-17RE is the functional receptor for IL-17C and mediates mucosal immunity to infection with intestinal pathogens.
Nature immunology (2011-10-14)
Xinyang Song, Shu Zhu, Peiqing Shi, Yan Liu, Yufang Shi, Steven D Levin, Youcun Qian
ABSTRACT
Interleukin 17 receptor E (IL-17RE) is an orphan receptor of the IL-17 receptor family. Here we show that IL-17RE is a receptor specific to IL-17C and has an essential role in host mucosal defense against infection. IL-17C activated downstream signaling through IL-17RE-IL-17RA complex for the induction of genes encoding antibacterial peptides as well as proinflammatory molecules. IL-17C was upregulated in colon epithelial cells during infection with Citrobacter rodentium and acted in synergy with IL-22 to induce the expression of antibacterial peptides in colon epithelial cells. Loss of IL-17C-mediated signaling in IL-17RE-deficient mice led to lower expression of genes encoding antibacterial molecules, greater bacterial burden and early mortality during infection. Together our data identify IL-17RE as a receptor of IL-17C that regulates early innate immunity to intestinal pathogens.