MilliporeSigma
  • Mild phenotype of epidermolytic hyperkeratosis mimicking ichthyosis bullosa of Siemens is related to specific mutation in 2B domain of KRT1.
Mild phenotype of epidermolytic hyperkeratosis mimicking ichthyosis bullosa of Siemens is related to specific mutation in 2B domain of KRT1.
Journal of dermatological science (2013-04-30)
Jae Yong Sung, Se-Woong Oh, Song Ee Kim, Soo-Chan Kim