MilliporeSigma
  • The mechanism of the cationic cyclooligomerization of ethylene-oxide and the concomitant degradation of the crown ether products formed.
The mechanism of the cationic cyclooligomerization of ethylene-oxide and the concomitant degradation of the crown ether products formed.
Acta Chemica Scandinavica. Series B (1980)
Dale J and Daasvatn K.