MilliporeSigma
  • STARD3 or STARD3NL and VAP form a novel molecular tether between late endosomes and the ER.
STARD3 or STARD3NL and VAP form a novel molecular tether between late endosomes and the ER.
Journal of cell science (2013-10-10)
Fabien Alpy, Adrien Rousseau, Yannick Schwab, François Legueux, Isabelle Stoll, Corinne Wendling, Coralie Spiegelhalter, Pascal Kessler, Carole Mathelin, Marie-Christine Rio, Timothy P Levine, Catherine Tomasetto
ABSTRACT
Inter-organelle membrane contacts sites (MCSs) are specific subcellular regions favoring the exchange of metabolites and information. We investigated the potential role of the late-endosomal membrane-anchored proteins StAR related lipid transfer domain-3 (STARD3) and STARD3 N-terminal like (STARD3NL) in the formation of MCSs involving late-endosomes (LEs). We demonstrate that both STARD3 and STARD3NL create MCSs between LEs and the endoplasmic reticulum (ER). STARD3 and STARD3NL use a conserved two phenylalanines in an acidic tract (FFAT)-motif to interact with ER-anchored VAP proteins. Together, they form an LE-ER tethering complex allowing heterologous membrane apposition. This LE-ER tethering complex affects organelle dynamics by altering the formation of endosomal tubules. An in situ proximity ligation assay between STARD3, STARD3NL and VAP proteins identified endogenous LE-ER MCS. Thus, we report here the identification of proteins involved in inter-organellar interaction.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution