MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A common variant in combination with a nonsense mutation in a member of the thioredoxin family causes primary ciliary dyskinesia.
A common variant in combination with a nonsense mutation in a member of the thioredoxin family causes primary ciliary dyskinesia.
Proceedings of the National Academy of Sciences of the United States of America (2007-03-16)
Bénédicte Duriez, Philippe Duquesnoy, Estelle Escudier, Anne-Marie Bridoux, Denise Escalier, Isabelle Rayet, Elisabeth Marcos, Anne-Marie Vojtek, Jean-François Bercher, Serge Amselem
ABSTRACT
Thioredoxins belong to a large family of enzymatic proteins that function as general protein disulfide reductases, therefore participating in several cellular processes via redox-mediated reactions. So far, none of the 18 members of this family has been involved in human pathology. Here we identified TXNDC3, which encodes a thioredoxin-nucleoside diphosphate kinase, as a gene implicated in primary ciliary dyskinesia (PCD), a genetic condition characterized by chronic respiratory tract infections, left-right asymmetry randomization, and male infertility. We show that the disease, which segregates as a recessive trait, results from the unusual combination of the following two transallelic defects: a nonsense mutation and a common intronic variant found in 1% of control chromosomes. This variant affects the ratio of two physiological TXNDC3 transcripts: the full-length isoform and a novel isoform, TXNDC3d7, carrying an in-frame deletion of exon 7. In vivo and in vitro expression data unveiled the physiological importance of TXNDC3d7 (whose expression was reduced in the patient) and the corresponding protein that was shown to bind microtubules. PCD is known to result from defects of the axoneme, an organelle common to respiratory cilia, embryonic nodal cilia, and sperm flagella, containing dynein arms, with, to date, the implication of genes encoding dynein proteins. Our findings, which identify a another class of molecules involved in PCD, disclose the key role of TXNDC3 in ciliary function; they also point to an unusual mechanism underlying a Mendelian disorder, which is an SNP-induced modification of the ratio of two physiological isoforms generated by alternative splicing.