MilliporeSigma
  • Kinetics and equilibrium studies from the methylene blue adsorption on diatomite treated with sodium hydroxide.
Kinetics and equilibrium studies from the methylene blue adsorption on diatomite treated with sodium hydroxide.
Applied Clay Science (2013)
Zhang J, et al.