MilliporeSigma
  • Matrilysin-2 expression in colorectal cancer is associated with overall survival of patients.
Matrilysin-2 expression in colorectal cancer is associated with overall survival of patients.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2013-12-10)
Qinglin Hu, Chuanjing Yan, Chengfei Xu, Hai Yan, Lian Qing, Yanjun Pu, Zeyou He, Xiaojun Li
ABSTRACT
Tumor recurrence and metastasis are pressing issues of patients with colorectal cancer who receive surgery. Matrilysin-2 (MMP-26) has been proved to play an important role during invasion and metastasis of some human solid tumor. We aimed to investigate the clinical significance and prognostic value of matrilysin-2 in human colorectal cancer. Colorectal cancer and adjacent normal samples from 201 patients were collected. Matrilysin-2 expression level was investigated by immunohistochemistry assay, and its association with overall survival of patients was analyzed by statistical analysis. Results showed that matrilysin-2 expression level significantly elevated in colorectal cancer compared with adjacent normal specimens. Matrilysin-2 expression was also found to be associated with cancer invasion, lymph node metastasis, distant metastasis, and TNM stage. In addition, survival analysis showed that elevated matrilysin-2 expression was associated with poor overall survival of patients. Cox's proportional hazards model indicated that matrilysin-2 was an independent prognostic marker for patients with colorectal cancer. The present study found that the expression of matrilysin-2 increased in colorectal cancer and was associated with tumor progression. It also provided the first evidence that matrilysin-2 expression was an independent prognostic factor for patients with colorectal cancer, which might be a high specific biomarker for colorectal cancer.