MilliporeSigma
  • Potassium-promoted Mo/MWCNTs catalysts for effective partial oxidation of 1-dodecanol to 1-dodecanal.
Potassium-promoted Mo/MWCNTs catalysts for effective partial oxidation of 1-dodecanol to 1-dodecanal.
Applied Catalysis A: General (2015)
Liao SY and Ko AN.