MilliporeSigma
  • siRNA knockdown of mitochondrial thymidine kinase 2 (TK2) sensitizes human tumor cells to gemcitabine.
siRNA knockdown of mitochondrial thymidine kinase 2 (TK2) sensitizes human tumor cells to gemcitabine.
Oncotarget (2015-06-19)
Christine Di Cresce, Rene Figueredo, Mateusz Rytelewski, Saman Maleki Vareki, Colin Way, Peter J Ferguson, Mark D Vincent, James Koropatnick
ABSTRACT
Nucleoside metabolism enzymes are determinants of chemotherapeutic drug activity. The nucleoside salvage enzyme deoxycytidine kinase (dCK) activates gemcitabine (2', 2'-difluoro-2'-deoxycytidine) and is negatively regulated by deoxycytidine triphosphate (dCTP). Reduction of dCTP in tumor cells could, therefore, enhance gemcitabine activity. Mitochondrial thymidine kinase 2 (TK2) phosphorylates deoxycytidine to generate dCTP. We hypothesized that: (1) TK2 modulates human tumor cell sensitivity to gemcitabine, and (2) antisense knockdown of TK2 would decrease dCTP and increase dCK activity and gemcitabine activation. siRNA downregulation of TK2 sensitized MCF7 and HeLa cells (high and moderate TK2) but not A549 cells (low TK2) to gemcitabine. Combined treatment with TK2 siRNA and gemcitabine increased dCK. We also hypothesized that TK2 siRNA-induced drug sensitization results in mitochondrial damage that enhances gemcitabine effectiveness. TK2 siRNA and gemcitabine decreased mitochondrial redox status, DNA content, and activity. This is the first demonstration of a direct role for TK2 in gemcitabine resistance, or any independent role in cancer drug resistance, and further distinguishes TK2 function from that of other dTMP-producing enzymes [cytosolic TK1 and thymidylate synthase (TS)]. siRNA knockdown of TK1 and/or TS did not sensitize cancer cells to gemcitabine indicating that, among the 3 enzymes, only TK2 is a candidate therapeutic target for combination with gemcitabine.
Sigma-Aldrich
Anti-TK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.