MilliporeSigma
  • Voltammetric Analysis on the Formation of Fe (OH)2 and FeCO3, and on the Reactivity of Passivation of Steel in Carbonate Solutions.
Voltammetric Analysis on the Formation of Fe (OH)<SUB>2</SUB> and FeCO<SUB>3</SUB>, and on the Reactivity of Passivation of Steel in Carbonate Solutions.
Journal of Materials Engineering and Performance (2015)
Eliyan FF, et al.