MilliporeSigma
  • Matrix-assisted laser desorption/ionization time-of-flight vs. fast-atom bombardment and electrospray ionization mass spectrometry in the structural characterization of bacterial poly(3-hydroxyalkanoates).
Matrix-assisted laser desorption/ionization time-of-flight vs. fast-atom bombardment and electrospray ionization mass spectrometry in the structural characterization of bacterial poly(3-hydroxyalkanoates).
Rapid communications in mass spectrometry : RCM (2015-09-18)
Giuseppe Impallomeni, Alberto Ballistreri, Giovanni Marco Carnemolla, Domenico Franco, Salvatore P P Guglielmino
ABSTRACT
Bacterial poly(3-hydroxyalkanoates) (PHAs) are an emergent class of plastic materials available from renewable resources. Their properties are strictly correlated with the comonomeric composition and sequence, which may be determined by various mass spectrometry approaches. In this paper we compare fast-atom bombardment (FAB) and electrospray ionization (ESI) to matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS) of partially pyrolyzed samples. We determined the compositions and sequences of the medium-chain-length PHAs (mcl-PHAs) prepared by bacterial fermentation of Pseudomonas aeruginosa ATCC 27853 cultured in media containing fatty acids with 8, 12, 14, 18, and 20 carbon atoms as carbon sources by means of MALDI-TOFMS of pyrolyzates, and compared the results with those obtained by FAB- and ESI-MS in previous studies. MALDI matrices used were 9-aminoacridine (9-AA) and indoleacrylic acid (IAA). MALDI-TOFMS was carried out in negative ion mode when using 9-AA as a matrix, giving a semi-quantitative estimation of the 3-hydroxyacids constituting the PHAs, and in positive mode when using IAA, allowing us, through statistical analysis of the relative intensity of the oligomers generated by pyrolysis, to establish that the polymers obtained are true random copolyesters and not a mixture of homopolymers or copolymers. MALDI-TOFMS in 9-AA and IAA of partial pyrolyzates of mcl-PHAs represents a powerful method for the structural analysis of these materials. In comparison with FAB and ESI, MALDI provided an extended mass range with better sensitivity at higher mass and a faster method of analysis.
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free