MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Assessing tumor cytoarchitecture using multiecho DSC-MRI derived measures of the transverse relaxivity at tracer equilibrium (TRATE).
Assessing tumor cytoarchitecture using multiecho DSC-MRI derived measures of the transverse relaxivity at tracer equilibrium (TRATE).
Magnetic resonance in medicine (2014-09-18)
Natenael B Semmineh, Junzhong Xu, Jack T Skinner, Jingping Xie, Hua Li, Gregory Ayers, C Chad Quarles
ABSTRACT
In brain tumor dynamic susceptibility contrast (DSC)-MRI studies, multiecho acquisition methods are used to quantify the dynamic changes in T1 and T2 * that occur when contrast agent (CA) extravasates. Such methods also enable the estimation of the effective tissue CA transverse relaxivity. The goal of this study was to evaluate the sensitivity of the transverse relaxivity at tracer equilibrium (TRATE) to tumor cytoarchitecture. Computational and in vitro studies were used to evaluate the biophysical basis of TRATE. In 9L, C6, and human brain tumors, TRATE, the apparent diffusion coefficient (ADC), the CA transfer constant (K(trans) ), the extravascular extracellular volume fraction (ve ), and histological data were compared. Simulations and in vitro results indicate that TRATE is highly sensitive to variations in cellular properties such as cell size and density. The histologic cell density and TRATE values were significantly higher in 9L tumors as compared to C6 tumors. In animal and human tumors, a voxel-wise comparison of TRATE with ADC, ve , and K(trans) maps showed low spatial correlation. The assessment of TRATE is clinically feasible and its sensitivity to tissue cytoarchitectural features not present in other imaging methods indicate that it could potentially serve as a unique structural signature or "trait" of cancer.
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)