MilliporeSigma
  • Tumour antigen expression in hepatocellular carcinoma in a low-endemic western area.
Tumour antigen expression in hepatocellular carcinoma in a low-endemic western area.
British journal of cancer (2015-06-10)
K Sideras, S J Bots, K Biermann, D Sprengers, W G Polak, J N M IJzermans, R A de Man, Q Pan, S Sleijfer, M J Bruno, J Kwekkeboom
ABSTRACT
Identification of tumour antigens is crucial for the development of vaccination strategies against hepatocellular carcinoma (HCC). Most studies come from eastern-Asia, where hepatitis-B is the main cause of HCC. However, tumour antigen expression is poorly studied in low-endemic, western areas where the aetiology of HCC differs. We constructed tissue microarrays from resected HCC tissue of 133 patients. Expression of a comprehensive panel of cancer-testis (MAGE-A1, MAGE-A3/4, MAGE-A10, MAGE-C1, MAGE-C2, NY-ESO-1, SSX-2, sperm protein 17), onco-fetal (AFP, Glypican-3) and overexpressed tumour antigens (Annexin-A2, Wilms tumor-1, Survivin, Midkine, MUC-1) was determined by immunohistochemistry. A higher prevalence of MAGE antigens was observed in patients with hepatitis-B. Patients with expression of more tumour antigens in general had better HCC-specific survival (P=0.022). The four tumour antigens with high expression in HCC and no, or weak, expression in surrounding tumour-free-liver tissue, were Annexin-A2, GPC-3, MAGE-C1 and MAGE-C2, expressed in 90, 39, 17 and 20% of HCCs, respectively. Ninety-five percent of HCCs expressed at least one of these four tumour antigens. Interestingly, GPC-3 was associated with SALL-4 expression (P=0.001), an oncofetal transcription factor highly expressed in embryonal stem cells. SALL-4 and GPC-3 expression levels were correlated with vascular invasion, poor differentiation and higher AFP levels before surgery. Moreover, patients who co-expressed higher levels of both GPC-3 and SALL-4 had worse HCC-specific survival (P=0.018). We describe a panel of four tumour antigens with excellent coverage and good tumour specificity in a western area, low-endemic for hepatitis-B. The association between GPC-3 and SALL-4 is a novel finding and suggests that GPC-3 targeting may specifically attack the tumour stem-cell compartment.