MilliporeSigma
  • Human exposure to heavy metals and possible public health risks via consumption of wild edible mushrooms from Slovak Paradise National Park, Slovakia.
Human exposure to heavy metals and possible public health risks via consumption of wild edible mushrooms from Slovak Paradise National Park, Slovakia.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2015-09-12)
Július Árvay, Ján Tomáš, Martin Hauptvogl, Peter Massányi, Ľuboš Harangozo, Tomáš Tóth, Radovan Stanovič, Štefánia Bryndzová, Monika Bumbalová
ABSTRACT
The contamination level of 92 samples (12 species) of wild edible mushrooms and underlying substrates with heavy metals (Cd, Cu, Hg, Pb and Zn) in the Slovak Paradise National Park that borders with a region of historical mining and processing of polymetallic ores, were determined. The collected samples were analyzed using of atomic absorption spectrophotometry. The metals were determined separately in hymenophore (H) and rest of fruit bodies (RFB). Bioaccumulation factor as well as ratio of metal content in H and RFB were calculated. Cadmium and lead contents in hymenophore exceeded statutory limits of the EU (Cd: 0.5 mg/kg dry weight (dw), Pb: 1.0 mg/kg dw) for edible mushrooms in 96% and 83% of the samples, respectively. The risk from the consumption of the collected mushroom species was calculated based on the provisionally tolerable weekly intake (PTWI) values, and the highest health risk arising with consumption of particularly Macrolepiota procera, Marasmius oreades and Russula vesca from the observed area was demonstrated. It was shown that average weekly consumption of tested mushrooms species results the threat of exceeding of PTWI limits in the case of cadmium values (by 164%, 86% and 4% of PTWI for M. oreades, R. vesca and R. puellaris, respectively) and of mercury (by 96% of PTWI for M. procera) but not lead.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, SAJ first grade, 65.0-66.0%, density: 1.40
Sigma-Aldrich
Nitric acid, 0.1 M