MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Lipoxygenase-inhibiting phenolic glycosides and monoterpene glycosides from Paeonia lactiflora.
Lipoxygenase-inhibiting phenolic glycosides and monoterpene glycosides from Paeonia lactiflora.
Journal of Asian natural products research (2015-03-24)
Liang Zou, Lin-Feng Hu, Yi-Dong Guo, Yu Song, Qiang Fu
ABSTRACT
The EtOH extract of the roots of Paeonia lactiflora afforded a new phenolic glycoside paenoside A (1) and a new monoterpene glycoside paeonin D (2), and five known monoterpene glycosides. Their structures were elucidated on the basis of spectroscopic means and hydrolysis products. All compounds displayed inhibitory potential against enzyme lipoxygenase.
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture