MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Matrix-assisted laser desorption/ionization-time of flight mass spectrometry identification of large colony beta-hemolytic streptococci containing Lancefield groups A, C, and G.
Matrix-assisted laser desorption/ionization-time of flight mass spectrometry identification of large colony beta-hemolytic streptococci containing Lancefield groups A, C, and G.
Infectious diseases (London, England) (2015-05-12)
Christian Salgård Jensen, Casper Dam-Nielsen, Magnus Arpi
ABSTRACT
The aim of this study was to investigate whether large colony beta-hemolytic streptococci containing Lancefield groups A, C, and G can be adequately identified using matrix-assisted laser desorption/ionization-time of flight mass spectrometry (MALDI-ToF). Previous studies show varying results, with an identification rate from below 50% to 100%. Large colony beta-hemolytic streptococci containing Lancefield groups A, C, and G isolated from blood cultures between January 1, 2007 and May 1, 2012 were included in the study. Isolates were identified to the species level using a combination of phenotypic characteristics and 16s rRNA sequencing. The isolates were subjected to MALDI-ToF analysis. We used a two-stage approach starting with the direct method. If no valid result was obtained we proceeded to an extraction protocol. Scores above 2 were considered valid identification at the species level. A total of 97 Streptococcus pyogenes, 133 Streptococcus dysgalactiae, and 2 Streptococcus canis isolates were tested; 94%, 66%, and 100% of S. pyogenes, S. dysgalactiae, and S. canis, respectively, were correctly identified by MALDI-ToF. In most instances when the isolates were not identified by MALDI-ToF this was because MALDI-ToF was unable to differentiate between S. pyogenes and S. dysgalactiae. By removing two S. pyogenes reference spectra from the MALDI-ToF database the proportion of correctly identified isolates increased to 96% overall. MALDI-ToF is a promising method for discriminating between S. dysgalactiae, S. canis, and S. equi, although more strains need to be tested to clarify this.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%