MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Differential effects of polyoma virus middle tumor antigen mutants upon gap junctional, intercellular communication.
Differential effects of polyoma virus middle tumor antigen mutants upon gap junctional, intercellular communication.
Experimental cell research (2015-07-19)
Mulu Geletu, Stephanie Guy, Samantha Greer, Leda Raptis
ABSTRACT
Gap junctions are channels that connect the cytoplasm of adjacent cells. Oncogenes such as the middle Tumor antigen of polyoma virus (mT) are known to suppress gap junctional, intercellular communication (GJIC). mT associates with and is tyrosine-phosphorylated by cSrc family members. Specific mT phosphotyrosines provide docking sites for the phosphotyrosine binding domain of Shc (mT-tyr250) or the SH2 domain of the regulatory subunit of the phosphatidylinositol-3 kinase (PI3k, mT-tyr315). Binding results in the activation of their downstream signaling cascades, Ras/Raf/Erk and PI3 kinase/Akt, respectively, both of which are needed for full neoplastic transformation. To examine the effect of mT-initiated pathways upon gap junctional communication, GJIC was quantitated in rat liver epithelial T51B cells expressing mT-mutants, using a novel technique of in situ electroporation. The results demonstrate for the first time that, although even low levels of wild-type mT are sufficient to interrupt gap junctional communication, GJIC suppression still requires an intact tyr-250 site, that is activation of the Ras pathway. In sharp contrast, activation of the PI3k pathway is not required for GJIC suppression, indicating that GJIC suppression is independent of full neoplastic conversion and the concomitant morphological changes. Interestingly, expression of a constitutively active, myristylated form of the catalytic subunit of PI3k, p110, or the constitutively active mutants E545K and H1047R increased GJIC, while pharmacological inhibition of PI3k eliminated communication. Therefore, although PI3k is growth promoting and in an activated form it can act as an oncogene, it actually plays a positive role upon gap junctional, intercellular communication.
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
Bicinchoninic Acid solution