MilliporeSigma
  • SOD3 Variant, R213G, Altered SOD3 Function, Leading to ROS-Mediated Inflammation and Damage in Multiple Organs of Premature Aging Mice.
SOD3 Variant, R213G, Altered SOD3 Function, Leading to ROS-Mediated Inflammation and Damage in Multiple Organs of Premature Aging Mice.
Antioxidants & redox signaling (2015-05-01)
Myung-Ja Kwon, Kyo-Young Lee, Han-Woong Lee, Jung-Ho Kim, Tae-Yoon Kim
ABSTRACT
Among the isoforms of superoxide dismutase, SOD3 is uniquely associated with the extracellular matrix (ECM) by virtue of its heparin-binding domain (HBD). Substitution of arginine by glycine at amino acid 213 (R213G) of its HBD was first identified in patients with heart failure, followed by many studies that focused on the role of this variant (SOD3(R213G)) in ischemic heart disease and cardiovascular disease. However, the biological significance of this mutation in a physiological context is largely unknown. As a first step, we generated SOD3(R213G) transgenic mice, in which the variant gene was driven by the β-actin promoter allowing expression in all tissues. Unexpectedly, we found that SOD3(R213G) transgenic mice exhibited premature aging, including hair graying, abnormal gait, and a shortened life span. Specifically, the aged mice showed systemic inflammation and organ degeneration. In addition, aged SOD3(R213G) mice are susceptible to neutrophil-mediated inflammation. Among other functions, the neutrophils of SOD3(R213G) mice produce high amounts of reactive oxygen species, which would normally be controlled by SOD3 in ECM. These findings showed for the first time that arginine 213 in the HBD of SOD3 is critical for maintaining proper organ function through moderating the normal innate immune response, which would otherwise lead to chronic inflammation and degenerative diseases in aged mice. Therefore, patients with this variant may be treated with SOD3 as a therapeutic strategy to prevent or cure these diseases.
Sigma-Aldrich
DAPI, for nucleic acid staining