MilliporeSigma
  • Fatty Acids Composition of Vegetable Oils and Its Contribution to Dietary Energy Intake and Dependence of Cardiovascular Mortality on Dietary Intake of Fatty Acids.
Fatty Acids Composition of Vegetable Oils and Its Contribution to Dietary Energy Intake and Dependence of Cardiovascular Mortality on Dietary Intake of Fatty Acids.
International journal of molecular sciences (2015-06-10)
Jana Orsavova, Ladislava Misurcova, Jarmila Vavra Ambrozova, Robert Vicha, Jiri Mlcek
ABSTRACT
Characterizations of fatty acids composition in % of total methylester of fatty acids (FAMEs) of fourteen vegetable oils--safflower, grape, silybum marianum, hemp, sunflower, wheat germ, pumpkin seed, sesame, rice bran, almond, rapeseed, peanut, olive, and coconut oil--were obtained by using gas chromatography (GC). Saturated (SFA), monounsaturated (MUFA) and polyunsaturated fatty acids (PUFA), palmitic acid (C16:0; 4.6%-20.0%), oleic acid (C18:1; 6.2%-71.1%) and linoleic acid (C18:2; 1.6%-79%), respectively, were found predominant. The nutritional aspect of analyzed oils was evaluated by determination of the energy contribution of SFAs (19.4%-695.7% E(RDI)), PUFAs (10.6%-786.8% E(RDI)), n-3 FAs (4.4%-117.1% E(RDI)) and n-6 FAs (1.8%-959.2% E(RDI)), expressed in % E(RDI) of 1 g oil to energy recommended dietary intakes (E(RDI)) for total fat (E(RDI)--37.7 kJ/g). The significant relationship between the reported data of total fat, SFAs, MUFAs and PUFAs intakes (% E(RDI)) for adults and mortality caused by coronary heart diseases (CHD) and cardiovascular diseases (CVD) in twelve countries has not been confirmed by Spearman's correlations.
Sigma-Aldrich
Methyl undecanoate, ≥98.0% (GC)
Pricing and availability is not currently available.