MilliporeSigma
  • A new class of bradykinin antagonists: synthesis and in vitro activity of bissuccinimidoalkane peptide dimers.
A new class of bradykinin antagonists: synthesis and in vitro activity of bissuccinimidoalkane peptide dimers.
Journal of medicinal chemistry (1992-05-01)
J C Cheronis, E T Whalley, K T Nguyen, S R Eubanks, L G Allen, M J Duggan, S D Loy, K A Bonham, J K Blodgett
ABSTRACT
A systematic study on the dimerization of the bradykinin (BK) antagonist D-Arg0-Arg1-Pro2-Hyp3-Gly4-Phe5-Ser6-D-Phe 7-Leu8-Arg9 has been performed. The first part of this study involved compounds wherein dimerization was carried out by sequentially replacing each amino acid with cysteine and cross-linking with bismaleimidohexane. The second part of this study utilized a series of bissuccinimidoalkane dimers wherein the intervening methylene chain was varied systematically from n = 2 to n = 12 while the point of dimerization was held constant at position 6. The biological activities of these dimers were then evaluated on BK-induced smooth muscle contraction in two different isolated tissue preparations: guinea pig ileum (GPI) and rat uterus (RU). Several of the dimeric BK antagonists displayed remarkable activities and long durations of action. In addition, dimerization at position 4, 7, 8, or 9 produced dimeric analogues with markedly reduced potency. Rank order of antagonist potency as a function of dimerization position is as follows: rat uterus, 6 greater than 5 greater than 0 greater than 2 greater than 1 greater than 3 much greater than 4, 7, 8, 9; guinea pig ileum, 6 greater than 5 greater than 3 greater than 2 greater than 1 greater than 0 much greater than 4, 7, 8, 9. Evaluation of the linker length as represented by the number of methylene units indicated an optimal distance between the two monomeric peptides of six to eight methylene moieties. These studies also revealed that the carbon-chain length significantly affected the duration of action in vitro and resulted in partial agonism effects when n greater than 8. The optimum activity in vitro was achieved with dimerization at position 6 and n = 6 (designated herein as compound 25; alternatively, CP-0127). Similar effects in potency were also seen when the monomeric antagonist D-Arg0-Arg1-Pro2-Hyp3-Gly4-Phe5-Ser6-D-Phe 7-Phe8-Arg9 (NPC-567) was dimerized using similar chemistry. These results suggest that the development of BK antagonists of significant therapeutic potential may be possible using a dimerization strategy that can overcome the heretofore limiting problems of potency and in vivo duration of action found with many of the BK antagonists in the literature.