MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Two new cucurbitane-type triterpenoid saponins isolated from ethyl acetate extract of Citrullus colocynthis fruit.
Two new cucurbitane-type triterpenoid saponins isolated from ethyl acetate extract of Citrullus colocynthis fruit.
Journal of Asian natural products research (2015-03-12)
Fei Song, Bin Dai, Hai-Yan Zhang, Jian-Wei Xie, Cheng-Zhi Gu, Jie Zhang
ABSTRACT
Two new cucurbitacins I (1 and 2), together with eight known compounds (3-10), were isolated from the ethyl acetate extract of the fruit of Citrullus colocynthis. Compounds 3, 5-9 were isolated from C. colocynthis for the first time. The structures of new compounds were determined primarily from IR, HR-MS, 1D-, and 2D-NMR analysis.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%