MilliporeSigma
  • Butein, a plant polyphenol, induces apoptosis concomitant with increased caspase-3 activity, decreased Bcl-2 expression and increased Bax expression in HL-60 cells.
Butein, a plant polyphenol, induces apoptosis concomitant with increased caspase-3 activity, decreased Bcl-2 expression and increased Bax expression in HL-60 cells.
Pharmacology & toxicology (2001-06-08)
N Y Kim, H O Pae, G S Oh, T H Kang, Y C Kim, H Y Rhew, H T Chung
ABSTRACT
In the present study we have investigated whether butein could induce apoptosis in human leukaemic HL-60 cells. The treatment of HL-60 cells with butein induced apoptotic cell death as determined by morphological and biochemical changes. Apoptotic DNA fragments in the butein-treated HL-60 cells were increased gradually as determined by flow cytometric analysis. The caspase-3 activity was increased during butein-induced apoptosis. However, caspase-3 inhibitor abrogated the butein-induced DNA fragmentation. Furthermore, the treatment of HL-60 cells with butein decreased the expression of Bcl-2 protein, but increased the expression of Bax protein. These results suggest that butein-induced apoptosis is mediated through the activation of caspase-3 and it is associated with changed expression of Bcl-2 and Bax proteins.