MilliporeSigma
  • Expanded scope in ethylene-alkyne cross-metathesis: coordinating heteroatom functionality at the propargylic position.
Expanded scope in ethylene-alkyne cross-metathesis: coordinating heteroatom functionality at the propargylic position.
Organic letters (2000-08-10)
J A Smulik, S T Diver
ABSTRACT
The Grubbs 1,3-dimesityl-4,5-dihydroimidazol-2-ylidene-substituted ruthenium complex 1 catalyzed ethylene-alkyne cross-metathesis and was shown to tolerate free hydroxyl groups and coordinating functionality at the propargylic and homopropargylic positions. Hindered and enantiomerically enriched 1-substituted alkynes also react efficiently under the reported conditions.
Sigma-Aldrich
Grubbs Catalyst® M204, Umicore