MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of environmental hypercapnia and metal (Cd and Cu) exposure on acid-base and metal homeostasis of marine bivalves.
Effects of environmental hypercapnia and metal (Cd and Cu) exposure on acid-base and metal homeostasis of marine bivalves.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2015-05-27)
Anna V Ivanina, Chelsea Hawkins, Elia Beniash, Inna M Sokolova
ABSTRACT
Elevated CO2 levels reduce seawater pH and may affect bioavailability of trace metals in estuaries. We studied the interactive effects of common metal pollutants (50 μg l(-1) Cd or Cu) and PCO2 (~395, 800 and 2000 μatm) on metal levels, intracellular pH, expression of metal binding proteins and stress biomarkers in estuarine bivalves Crassostrea virginica (oysters) and Mercenaria mercenaria (hard clams). Cd (but not Cu or hypercapnia) exposure affected the acid-base balance of hemocytes resulting in elevated intracellular pH. Cd and Cu exposure led to the increase in the tissue metal burdens, and metal accumulation was reduced by elevated PCO2 in the mantle but not hemocytes. No change was found in the intracellular free Cd(2+), Cu(2+) or Fe(2+) during Cu or Cd exposure indicating that these metals are bound to intracellular ligands. Free Zn(2+) content in oyster hemocytes was suppressed by Cd and Cu exposure and below the detection limits in clam hemocytes, which went hand-in-hand with the elevated mRNA expression of metallothioneins and ferritin in Cd- and Cu-exposed bivalves, enhanced by hypercapnia. The metal-binding and antioxidant mechanisms of oysters and clams were sufficient to effectively maintain intracellular redox status, even though metal exposure combined with moderate hypercapnia (~800 μatm PCO2) led to the elevated production of reactive oxygen species in hemocytes. Overall, while hypercapnia modulates metal accumulation, binding capacity and oxidative stress in estuarine bivalves, the physiological effects of elevated CO2 are mild compared to the effects of other common stressors.
Sigma-Aldrich
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein, ≥90% (HPLC)
Sigma-Aldrich
N-Bromosuccinimide, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)