MilliporeSigma
  • Ombitasvir plus paritaprevir plus ritonavir with or without ribavirin in treatment-naive and treatment-experienced patients with genotype 4 chronic hepatitis C virus infection (PEARL-I): a randomised, open-label trial.
Ombitasvir plus paritaprevir plus ritonavir with or without ribavirin in treatment-naive and treatment-experienced patients with genotype 4 chronic hepatitis C virus infection (PEARL-I): a randomised, open-label trial.
Lancet (London, England) (2015-04-04)
Christophe Hézode, Tarik Asselah, K Rajender Reddy, Tarek Hassanein, Marina Berenguer, Katarzyna Fleischer-Stepniewska, Patrick Marcellin, Coleen Hall, Gretja Schnell, Tami Pilot-Matias, Niloufar Mobashery, Rebecca Redman, Regis A Vilchez, Stanislas Pol
ABSTRACT
Hepatitis C virus (HCV) genotype 4 accounts for about 13% of global HCV infections. Because interferon-containing treatments for genotype 4 infection have low efficacy and poor tolerability, an unmet need exists for effective all-oral regimens. We examined the efficacy and safety of an all-oral interferon-free regimen of ombitasvir, an NS5A inhibitor, and paritaprevir (ABT-450), an NS3/4A protease inhibitor dosed with ritonavir (ombitasvir plus paritaprevir plus ritonavir), given with or without ribavirin. In this multicentre ongoing phase 2b, randomised, open-label combination trial (PEARL-I), patients were recruited from academic, public, and private hospitals and clinics in France, Hungary, Italy, Poland, Romania, Spain, Turkey, and the USA. Eligible participants were aged 18-70 years with non-cirrhotic, chronic HCV genotype 4 infection (documented ≥6 months before screening) and plasma HCV RNA levels higher than 10,000 IU/mL. Previously untreated (treatment-naive) patients were randomly assigned (1:1) by computer-generated randomisation lists to receive once-daily ombitasvir (25 mg) plus paritaprevir (150 mg) plus ritonavir (100 mg) with or without weight-based ribavirin for 12 weeks. Previously treated (treatment-experienced) patients who had received pegylated interferon plus ribavirin all received the ribavirin-containing regimen. The primary endpoint was a sustained virological response (HCV RNA <25 IU/mL) 12 weeks after the end of treatment (SVR12). Analysis was by intention to treat. This study is registered with ClinicalTrials.gov, number NCT01685203. Between Aug 14, 2012, and Nov 19, 2013, 467 patients with HCV infection were screened, of whom 174 were infected with genotype 4. 135 patients were randomly assigned to treatment and received at least one dose of study medication; 86 patients were treatment-naive, of whom 44 received ombitasvir plus paritaprevir plus ritonavir and 42 received ombitasvir plus paritaprevir plus ritonavir with ribavirin, and 49 treatment-experienced patients received the ribavirin-containing regimen. In previously untreated patients, SVR12 rates were 100% (42/42 [95% CI 91·6-100]) in the ribavirin-containing regimen and 90·9% (40/44 [95% CI 78·3-97·5]) in the ribavirin-free regimen. No statistically significant differences in SVR12 rates were noted between the treatment-naive groups (mean difference -9·16% [95% CI -19·61 to 1·29]; p=0·086). All treatment-experienced patients achieved SVR12 (49/49; 100% [95% CI 92·7-100]). In the ribavirin-free group, two (5%) of 42 treatment-naive patients had virological relapse, and one (2%) of 44 had virological breakthrough; no virological failures were recorded in the ribavirin-containing regimen. The most common adverse event was headache (14 [29%] of 49 treatment-experienced patients and 14 [33%] of 42 treatment-naive patients). No adverse event-related discontinuations or dose interruptions of study medications, including ribavirin, were noted, and only four patients (4%) of 91 receiving ribavirin required dose modification for haemoglobin less than 100 g/L or anaemia. An interferon-free regimen of ombitasvir plus paritaprevir plus ritonavir with or without ribavirin achieved high sustained virological response rates at 12 weeks after the end of treatment and was generally well tolerated, with low rates of anaemia and treatment discontinuation in non-cirrhotic previously untreated and previously treated patients with HCV genotype 4 infection. AbbVie.
Sigma-Aldrich
Ribavirin, antiviral