MilliporeSigma
  • Estimation of eye lens doses received by pediatric interventional cardiologists.
Estimation of eye lens doses received by pediatric interventional cardiologists.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2015-06-07)
L Alejo, C Koren, C Ferrer, E Corredoira, A Serrada
ABSTRACT
Maximum Hp(0.07) dose to the eye lens received in a year by the pediatric interventional cardiologists has been estimated. Optically stimulated luminescence dosimeters were placed on the eyes of an anthropomorphic phantom, whose position in the room simulates the most common irradiation conditions. Maximum workload was considered with data collected from procedures performed in the Hospital. None of the maximum values obtained exceed the dose limit of 20 mSv recommended by ICRP.
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Rhenium, wire reel, 2m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.125mm, annealed, 99.97%
Rhenium, wire reel, 1m, diameter 0.125mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Rhenium, wire reel, 2m, diameter 0.178mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 1m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Rhenium, wire reel, 1m, diameter 0.178mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.076mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Rhenium, wire reel, 1m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.2mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Rhenium, wire reel, 0.025m, diameter 1.0mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Rhenium, wire reel, 0.05m, diameter 0.5mm, annealed, 99.97%
Rhenium, wire reel, 2m, diameter 0.2mm, annealed, 99.97%