MilliporeSigma
  • Human Surfactant Proteins A2 (SP-A2) and B (SP-B) Genes as Determinants of Respiratory Distress Syndrome.
Human Surfactant Proteins A2 (SP-A2) and B (SP-B) Genes as Determinants of Respiratory Distress Syndrome.
Indian pediatrics (2015-06-11)
Walaa A Abuelhamed, Nancy Zeidan, Walaa A Shahin, Hoda I Rizk, Walaa A Rabie
ABSTRACT
To study the relationship between SP-A2 and SP-B gene polymorphisms and respiratory distress syndrome in preterm neonates. Cross-sectional. Neonatal intensive care unit and the Molecular Biology unit of the Chemical Pathology Department, Kasr Alainy hospital, Cairo University. Sixty-five preterm infants with respiratory distress syndrome and 50 controls. The genomic DNA was isolated using DNA extraction kits. SYBR Green-based real-time PCR was used to determine the variant genotypes of SP-A2 c.751 G>A and SP-B c.8714 G>C single nucleotide polymorphisms. Homozygosity of SP-A (OR 46, 95% CI 14-151) and SP-B (OR 5.2, 95% CI 2.3-11.4) alleles increased the risk of respiratory distress syndrome. The logistic regression model showed that genotypes SP-A2 (OR 164) and SP-B (OR 18) were directly related to the occurrence of respiratory distress syndrome, whereas gestational age (OR 0.57) and 5-minute Apgar score (OR 0.19) were inversely related to its occurrence. There is a possible involvement of SP-A2 and SP-B genes polymorphisms in the genetic predisposition to respiratory distress syndrome.