MilliporeSigma
  • The Use of Chemical-Chemical Interaction and Chemical Structure to Identify New Candidate Chemicals Related to Lung Cancer.
The Use of Chemical-Chemical Interaction and Chemical Structure to Identify New Candidate Chemicals Related to Lung Cancer.
PloS one (2015-06-06)
Lei Chen, Jing Yang, Mingyue Zheng, Xiangyin Kong, Tao Huang, Yu-Dong Cai
ABSTRACT
Lung cancer causes over one million deaths every year worldwide. However, prevention and treatment methods for this serious disease are limited. The identification of new chemicals related to lung cancer may aid in disease prevention and the design of more effective treatments. This study employed a weighted network, constructed using chemical-chemical interaction information, to identify new chemicals related to two types of lung cancer: non-small lung cancer and small-cell lung cancer. Then, a randomization test as well as chemical-chemical interaction and chemical structure information were utilized to make further selections. A final analysis of these new chemicals in the context of the current literature indicates that several chemicals are strongly linked to lung cancer.
Sigma-Aldrich
Lomustine, ≥98%
Sigma-Aldrich
Ifosfamide, ≥98%
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Indole, ≥99%