MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Promotion of liver and kidney carcinogenesis by ethyl tertiary-butyl ether (ETBE) in male Wistar rats.
Promotion of liver and kidney carcinogenesis by ethyl tertiary-butyl ether (ETBE) in male Wistar rats.
Journal of toxicologic pathology (2015-11-06)
Akihiro Hagiwara, Yuko Doi, Norio Imai, Mayuko Suguro, Mayumi Kawabe, Fumio Furukawa, Seiko Tamano, Kasuke Nagano, Shoji Fukushima
ABSTRACT
Tumor-promoting effects of ethyl tertiary-butyl ether (ETBE) were investigated in a 2-stage carcinogenesis bioassay with regard to hepatic and renal carcinogenesis in rats. Male 6-week-old Wistar rats were given drinking water containing N-ethyl-N-(2-hydroxyethyl)nitrosamine (EHEN), as an initiator, at a dose of 500 ppm for 2 weeks. Starting one week thereafter, the animals were administered ETBE at dose levels of 0 (control), 100, 300, 500 or 1,000 mg/kg/day by gavage for 19 weeks from week 4 to 22. Necropsy of all rats was performed at week 23, and livers and kidneys were examined histopathologically. Incidences of hepatocellular adenomas, and those of combined hepatocellular adenomas and carcinomas were significantly elevated in rats given 1,000 mg/kg/day ETBE, but not 100‒500 mg/kg/day ETBE, and there was a significant increase in the average numbers of lesions. No significant differences in incidences and average numbers of renal tubule neoplasms were found in rats administered 100‒1,000 mg/kg/day ETBE. However, the average numbers of atypical tubule hyperplasias, considered to be preneoplastic lesions, were significantly increased in rats given ETBE at 1,000 mg/kg/day, but not in rats given 500 mg/kg/day or lower doses. Thus, these results imply that ETBE has hepatic and renal tumor-promoting activities that affect EHEN-induced carcinogenesis in male rats, and the no-observed-effect level is 500 mg/kg/day under the present experimental conditions.
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)