MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Unmet needs associated with attention-deficit/hyperactivity disorder in eight European countries as reported by caregivers and adolescents: results from qualitative research.
Unmet needs associated with attention-deficit/hyperactivity disorder in eight European countries as reported by caregivers and adolescents: results from qualitative research.
The patient (2014-10-26)
Vanja Sikirica, Emuella Flood, C Noelle Dietrich, Javier Quintero, Val Harpin, Paul Hodgkins, Klaus Skrodzki, Kathleen Beusterien, M Haim Erder
ABSTRACT
Attention-deficit/hyperactivity disorder (ADHD) is a neurobehavioral disorder characterized by inattention, impulsivity, and hyperactivity, the levels of which are inappropriately high for an individual's developmental age. The objective of this study was to explore the unmet needs of children/adolescents with ADHD and their caregivers in eight European countries. Semi-structured interviews with 38 caregivers of children/adolescents (aged 6-17 years) with ADHD and no or less serious comorbidities and 28 adolescents (aged 13-17 years) with ADHD and no or less serious comorbidities were conducted, audio-recorded, transcribed into English, and coded for analysis. Caregivers reported their own ADHD-related issues, including making personal accommodations, such as limiting activities and spending extra time/effort caring for their child/adolescent, social impacts, and strained relationships. Medication was generally considered helpful; however, most children experience core ADHD symptoms while on medication (reported by 88 % of caregivers and 100 % of adolescents). Adolescents often reported schoolwork difficulties (96 %) and peer issues (75 %), while caregivers reported school issues (84 %) and peer difficulties (79 %). Caregivers reported minimal ADHD education and community support. Caregivers (29 %) and adolescents (54 %) desired medication that better controlled symptoms but had concerns about being oversubdued. Caregivers reported concerns about adverse effects (21 %). European caregivers of children/adolescents with ADHD identified multiple unmet needs, which persist despite treatment. Adolescents noted impacts on school and social interactions consistent with caregivers. Future research is needed to quantify the study findings, and, ultimately, ease the impact of ADHD on patients and their caregivers.
Sigma-Aldrich
(R)-Tomoxetine hydrochloride, solid