MilliporeSigma
  • Synthesis, characterization, and supercapacitor studies of manganese (IV) oxide nanowires.
Synthesis, characterization, and supercapacitor studies of manganese (IV) oxide nanowires.
Materials Science in Semiconductor Processing (2013)
Yousefi T, et al.