MilliporeSigma
Solid-phase synthesis of 1,7-disubstituted-1,3,5-triazepane-2,4-diones.
Organic letters (2001-09-01)
Y Yu, J M Ostresh, R A Houghten
ABSTRACT
[reaction: see text]. The solid-phase synthesis of 1,7-disubstituted-1,3,5-triazepane-2,4-diones from resin-bound amino acids is described. The exhaustive reduction of solid-support bound amides with borane afforded the requisite secondary amines, which following treatment with phenyl isocyanatoformate and cleavage, provided the corresponding triazepane-2,4-diones.