MilliporeSigma
  • Potent estrogen agonists based on carborane as a hydrophobic skeletal structure. A new medicinal application of boron clusters.
Potent estrogen agonists based on carborane as a hydrophobic skeletal structure. A new medicinal application of boron clusters.
Chemistry & biology (2001-04-28)
Y Endo, T Iijima, Y Yamakoshi, H Fukasawa, C Miyaura, M Inada, A Kubo, A Itai
ABSTRACT
Carboranes (dicarba-closo-dodecaboranes) are a class of carbon-containing polyhedral boron-cluster compounds having remarkable thermal stability and exceptional hydrophobicity. Applications of the unique structural and chemical properties offered by icosahedral carboranes in boron neutron capture therapy have received increasing attention over the past 30 years. However, these features of carboranes may allow another application as a hydrophobic pharmacophore in biologically active molecules that interact hydrophobically with receptors. We have designed candidate estrogen-receptor-binding compounds having carborane as a hydrophobic skeletal structure and synthesized them. The most potent compound bearing a carborane cage exhibited activity at least 10-fold greater than that of 17beta-estradiol in the luciferase reporter gene assay. Estrogen receptor-alpha-binding data for the compound were consistent with the results of the luciferase reporter gene assay. The compound also showed potent in vivo effects on the recovery of uterine weight and bone loss in ovariectomized mice. Further development of the potent carborane-containing estrogenic agonists described here, having a new skeletal structure and unique characteristics, should yield novel therapeutic agents, especially selective estrogen receptor modulators. Furthermore, the suitability of the spherical carborane cage for binding to the cavity of the estrogen receptor-alpha ligand-binding domain should provide a basis for a similar approach to developing novel ligands for other steroid receptors.