MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Platelet integrin alpha(IIb)beta(3): activation mechanisms.
Journal of thrombosis and haemostasis : JTH (2007-07-20)
Y-Q Ma, J Qin, E F Plow
ABSTRACT
Integrin alpha(IIb)beta(3) plays a critical role in platelet aggregation, a central response in hemostasis and thrombosis. This function of alpha(IIb)beta(3) depends upon a transition from a resting to an activated state such that it acquires the capacity to bind soluble ligands. Diverse platelet agonists alter the cytoplasmic domain of alpha(IIb)beta(3) and initiate a conformational change that traverses the transmembrane region and ultimately triggers rearrangements in the extracellular domain to permit ligand binding. The membrane-proximal regions of alpha(IIb) and beta(3) cytoplasmic tails, together with the transmembrane segments of the subunits, contact each other to form a complex which restrains the integrin in the resting state. It is unclasping of this complex that induces integrin activation. This clasping/unclasping process is influenced by multiple cytoplasmic tail binding partners. Among them, talin appears to be a critical trigger of alpha(IIb)beta(3) activation, but other binding partners, which function as activators or suppressors, are likely to act as co-regulators of integrin activation.