MilliporeSigma
  • Microphase Separation with Small Amphiphilic Molecules: Crystal Structure of Preservatives Sodium Benzoate (E 211) and Potassium Benzoate (E 212).
Microphase Separation with Small Amphiphilic Molecules: Crystal Structure of Preservatives Sodium Benzoate (E 211) and Potassium Benzoate (E 212).
Zeitschrift fur Anorganische und Allgemeine Chemie (2013)
Butterhof C, et al.