MilliporeSigma
  • NIPA1 gene mutations cause autosomal dominant hereditary spastic paraplegia (SPG6).
NIPA1 gene mutations cause autosomal dominant hereditary spastic paraplegia (SPG6).
American journal of human genetics (2003-09-26)
Shirley Rainier, Jing-Hua Chai, Debra Tokarz, Robert D Nicholls, John K Fink
ABSTRACT
The hereditary spastic paraplegias (HSPs) are genetically heterogeneous disorders characterized by progressive lower-extremity weakness and spasticity. The molecular pathogenesis is poorly understood. We report discovery of a dominant negative mutation in the NIPA1 gene in a kindred with autosomal dominant HSP (ADHSP), linked to chromosome 15q11-q13 (SPG6 locus); and precisely the same mutation in an unrelated kindred with ADHSP that was too small for meaningful linkage analysis. NIPA1 is highly expressed in neuronal tissues and encodes a putative membrane transporter or receptor. Identification of the NIPA1 function and ligand will aid an understanding of axonal neurodegeneration in HSP and may have important therapeutic implications.