MilliporeSigma
  • RaLP, a new member of the Src homology and collagen family, regulates cell migration and tumor growth of metastatic melanomas.
RaLP, a new member of the Src homology and collagen family, regulates cell migration and tumor growth of metastatic melanomas.
Cancer research (2007-04-06)
Ernesta Fagiani, Giuseppina Giardina, Lucilla Luzi, Matteo Cesaroni, Micaela Quarto, Maria Capra, Giovanni Germano, Maria Bono, Manuela Capillo, PierGiuseppe Pelicci, Luisa Lanfrancone
ABSTRACT
The Src homology and collagen (Src) family of adaptor proteins comprises six Shc-like proteins encoded by three loci in mammals (Shc, Rai, and Sli). Shc-like proteins are tyrosine kinase substrates, which regulate diverse signaling pathways and cellular functions, including Ras and proliferation (p52/p46Shc), phosphatidylinositol 3-kinase and survival (p54Rai), and mitochondrial permeability transition and apoptosis (p66Shc). Here, we report the identification, cloning, and sequence characterization of a new member of the Shc family that we termed RaLP. RaLP encodes a 69-kDa protein characterized by the CH2-PTB-CH1-SH2 modularity, typical of the Shc protein family, and expressed, among adult tissues, only in melanomas. Analysis of RaLP expression during the melanoma progression revealed low expression in normal melanocytes and benign nevi, whereas high levels of RaLP protein were found at the transition from radial growth phase to vertical growth phase and metastatic melanomas, when tumor cells acquire migratory competence and invasive potential. Notably, silencing of RaLP expression in metastatic melanomas by RNA interference reduced tumorigenesis in vivo. Analysis of RaLP in melanoma signal transduction pathways revealed that (a) when ectopically expressed in RaLP-negative melanocytes and nonmetastatic melanoma cells, it functions as a substrate of activated insulin-like growth factor-1 and epidermal growth factor receptors and increases Ras/mitogen-activated protein kinase (MAPK) signaling and cell migration, whereas (b) its silencing in RaLP-positive melanoma cells abrogates cell migration in vitro, without affecting MAPK signaling, suggesting that RaLP activates both Ras-dependent and Ras-independent migratory pathways in melanomas. These findings indicate that RaLP is a specific marker of metastatic melanomas, a critical determinant in the acquisition of the migratory phenotype by melanoma cells, and a potential target for novel anti-melanoma therapeutic strategies.