MilliporeSigma
(4-Methoxy-3-nitrobenzylideneamino)guanidinium chloride.
Acta crystallographica. Section C, Crystal structure communications (2007-07-05)
Joshua R Ring, Sean Parkin, Peter A Crooks
ABSTRACT
In the title compound, C(9)H(12)N(5)O(3)(+) x Cl(-), the cation is almost entirely planar. The imine double bond is exclusively in the E geometry.