MilliporeSigma
  • Chemopreventive effects of nobiletin and its colonic metabolites on colon carcinogenesis.
Chemopreventive effects of nobiletin and its colonic metabolites on colon carcinogenesis.
Molecular nutrition & food research (2015-10-08)
Xian Wu, Mingyue Song, Minqi Wang, Jinkai Zheng, Zili Gao, Fei Xu, Guodong Zhang, Hang Xiao
ABSTRACT
Nobiletin (NBT) is a major citrus flavonoid with various health benefits. Herein, we investigated the colon cancer chemopreventive effects of NBT and its colonic metabolites in a colitis-associated colon carcinogenesis mouse model as well as in human colon cancer cell models. In azoxymethane/dextran sulfate sodium treated mice, oral administration of NBT effectively decreased both incidence and multiplicity of colonic tumors. NBT showed significant antiproliferative, proapoptotic, and anti-inflammatory effects in the mouse colon. HPLC analysis revealed that oral administration of NBT resulted in high levels of metabolites, i.e. 3'-demethylnobiletin (M1), 4'-demethylnobiletin (M2), and 3',4'-didemethylnobiletin (M3) in the colonic mucosa. In contrast, the colonic level of NBT was about 20-fold lower than the total colonic level of three metabolites. Cell culture studies demonstrated that the colonic metabolites of NBT significantly inhibited the growth of human colon cancer cells, caused cell-cycle arrest, induced apoptosis, and profoundly modulated signaling proteins related with cell proliferation and cell death. All of these effects were much stronger than those produced by NBT alone. Our results demonstrated that oral administration of NBT significantly inhibited colitis-associated colon carcinogenesis in mice, and this chemopreventive effect was strongly associated with its colonic metabolites.
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%