MilliporeSigma
  • A benzenediamine derivative fc-99 attenuates lupus-like syndrome in MRL/lpr mice related to suppression of pDC activation.
A benzenediamine derivative fc-99 attenuates lupus-like syndrome in MRL/lpr mice related to suppression of pDC activation.
Immunology letters (2015-11-08)
Jianjian Ji, Hongye Fan, Fanlin Li, Xiaojing Li, Guanjun Dong, Wei Gong, Yuxian Song, Fei Liu, Chunyan Hua, Renxiang Tan, Huan Dou, Yayi Hou
ABSTRACT
Systemic lupus erythematosus (SLE) is an autoimmune disease with prominent chronic inflammatory aspects. Plasmacytoid dendritic cells (pDCs), which are the principal interferon-α (IFN-α)-producing cells, have known to be critically involved in SLE pathogenesis. Our previous research demonstrated that a benzenediamine derivative FC-99 possessed anti-inflammatory activities. However, the effects of FC-99 on SLE have not been investigated to date. In this study, we found that FC-99 attenuated lupus-like pathological symptoms and lupus nephritis as well as the expression of pro-inflammatory cytokines in kidneys of MRL/lpr mice. FC-99 also decreased both the total IgM, total IgG and anti-dsDNA IgG levels in sera and the activation of B cells in the PBMCs and spleens of MRL/lpr mice. Moreover, FC-99 inhibited the abnormal activation and number of pDCs from PBMCs and spleens and levels of IFN-α in MRL/lpr mice. Notably, FC-99 significantly suppressed the expression of IFN-inducible genes in peripheral blood mononuclear cells (PBMCs) and spleens from MRL/lpr mice. As expected, in vitro experiments demonstrated that FC-99 decreased both the activation and IFN-α production of pDCs and inhibited IRAK4 phosphorylation in pDCs upon TLR7 and TLR9 stimulation. We further confirm that the inhibition of FC-99 on B cell activation depended on level of pDCs-secreting IFN-α. These data indicate that FC-99 attenuated lupus-like syndrome in MRL/lpr mice related to suppression of pDC activation, especially pDCs-secreting IFN-α. This study suggests that FC-99 may be a potential therapeutic candidate for the treatment of SLE.
Sigma-Aldrich
FC-99 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)