MilliporeSigma
  • Comparative analysis of Pasteurella multocida strains isolated from bovine respiratory infections.
Comparative analysis of Pasteurella multocida strains isolated from bovine respiratory infections.
Acta microbiologica et immunologica Hungarica (2015-12-23)
Boglárka Sellyei, Zsuzsanna Rónai, Szilárd Jánosi, László Makrai
ABSTRACT
Bovine respiratory disease (BRD) is the leading cause of significant economic losses in the intensive beef industry worldwide. Beside numerous risk factors Pasteurella multocida, which is regarded as a secondary pathogen, may play a role in the development of the disease. Previous studies of strains from swine pneumonia revealed that there are a few clones associated with clinical disease, suggesting that some strains may be more virulent than others. This linkage may be true in the BRD, however composition of P. multocida populations in the herds are slightly characterized. Thus, we decided to perform phenotypic and genotypic characterisation of strains isolated from calves with respiratory infection at 31 different herds in Hungary. The results demonstrated the presence of two dominant strain types. At the identical taxonomic background (P. multocida subsp. multocida) with slight phenotypic variability they could be separated by trehalose fermentation capacity, α-glucosidase activity and molecular fingerprint patterns of ERIC- and M13-PCR. Independent prevalence and geographical origin of the strain types may refer to their significance in the illness, but their comparison with strains isolated from healthy individuals is taken into consideration.
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology