MilliporeSigma
  • Morphological and compositional monitoring of a new cell-free cartilage repair hydrogel technology - GelrinC by MR using semi-quantitative MOCART scoring and quantitative T2 index and new zonal T2 index calculation.
Morphological and compositional monitoring of a new cell-free cartilage repair hydrogel technology - GelrinC by MR using semi-quantitative MOCART scoring and quantitative T2 index and new zonal T2 index calculation.
Osteoarthritis and cartilage (2015-07-19)
S Trattnig, K Ohel, V Mlynarik, V Juras, S Zbyn, A Korner
ABSTRACT
To evaluate cartilage repair tissue (RT) using MOCART scoring for morphological and T2 mapping for biochemical assessment following implantation of GelrinC, a biosynthetic, biodegradable hydrogel implant. MR imaging (1.5/3T) was performed on 21 patients at six sites. Standard protocols were used for MOCART evaluation at 1 week (baseline) 1, 3, 6, 12, 18 and 24 months. Multi-echo SE was used for T2 mapping. Global (T2 in RT divided by T2 in normal cartilage) and zonal T2 index (deep T2 divided by superficial T2) of RT were calculated. Average MOCART score was 71.8 (95% CI 62.2 to 81.3) at six, 75.2 (95% CI 62.8 to 87.5) at twelve, 71.8 (95% CI 55.4 to 88.2) at eighteen and 84.4 (95% CI 77.7 to 91.0) at twenty-four months. The global T2 index ranged between 0.8 and 1.2 (normal healthy cartilage) in 1/11 (9%) patients at baseline, 8/12 (67%) at 12 months, 11/13 (85%) at 18 months and 13/16 (81%) at 24 months. The zonal T2 index for RT was <20% difference to the zonal T2 index for normal cartilage in: 6/12 patients (50%) at 12 months, 7/13 (53.8%) at 18 months and 10/16 (63.5%) at 24 months. The standard deviation for T2 showed a significant decrease over the study. The increase of MOCART scores over follow-up indicates improving cartilage repair tissue. Global and zonal T2 repair values at 24 months reached normal cartilage in 81% and 63.5% of the patients respectively, reflecting collagen organization similar to hyaline cartilage.
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)