MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The ability of the YAS and AR CALUX assays to detect the additive effects of anti-androgenic fungicide mixtures.
The ability of the YAS and AR CALUX assays to detect the additive effects of anti-androgenic fungicide mixtures.
Toxicology letters (2015-11-26)
Bettina Seeger, Frank Klawonn, Boris Nguema Bekale, Pablo Steinberg
ABSTRACT
Actual risk assessment only takes single pesticides into account, although about one third of the analyzed food samples in the European Union was contaminated with pesticide mixtures in 2013. A cumulative approach would group pesticides that share the same mechanism of toxicity. We evaluated the combination effects of low effect concentrations of binary and ternary mixtures of six anti-androgenic fungicides (procymidone, vinclozolin, tebuconazole, propiconazole, fenarimol and prochloraz) antagonizing the human androgen receptor (hAR) in the Yeast-based Androgen Screen assay (YAS) as well as in the AR Chemical-Activated LUciferase gene eXpression (AR CALUX) assay by means of concentration addition and nonlinear regression. The mixture effects were essentially additive when the fungicides were applied as iso-effective low inhibitory concentration combinations, independently from the used assay and as shown by the excellent agreement between experimental and predicted data. Both assays were successfully applied to evaluate the additive effects of fungicide mixtures at low concentrations. Since pesticide residues occur in/on foodstuffs in the EU at rather low concentrations and hormonally active environmental contaminants may concomitantly be present, more complex mixtures of anti-androgenic chemicals, not only pesticides, should be tested in the future when wanting to apply a target organ-based risk assessment approach.
Product Description
Supelco
Procymidone, PESTANAL®, analytical standard
Sigma-Aldrich
Hydroxyflutamide, ≥98% (HPLC)
Supelco
Vinclozolin, PESTANAL®, analytical standard