- A facile Er(OTf)3-catalyzed synthesis of 2,3-unsaturated O- and S-glycosides.
A facile Er(OTf)3-catalyzed synthesis of 2,3-unsaturated O- and S-glycosides.
Carbohydrate research (2007-06-23)
Antonio Procopio, Renato Dalpozzo, Antonio De Nino, Loredana Maiuolo, Monica Nardi, Manuela Oliverio, Beatrice Russo
PMID17583684
ABSTRACT
Er(OTf)(3) is a useful catalyst for the Ferrier rearrangement furnishing high yields of O- and S-glycosides. The transformation has wide applicability, cleaner reaction profiles, mild reaction conditions, and high stereoselectivity and the catalyst, which is also commercially available, can be recovered and reused.