MilliporeSigma
  • Ten-year efforts and a no-go recommendation for sodium borohydride for on-board automotive hydrogen storage
Ten-year efforts and a no-go recommendation for sodium borohydride for on-board automotive hydrogen storage
International Journal of Hydrogen Energy (2009)
Demirci UB, et al.