MilliporeSigma
  • Squaric acids: a new motif for designing inhibitors of protein tyrosine phosphatases.
Squaric acids: a new motif for designing inhibitors of protein tyrosine phosphatases.
Organic letters (2004-01-03)
Jian Xie, Anthony B Comeau, Christopher T Seto
ABSTRACT
[structure: see text] Protein tyrosine phosphatases (PTPases) are important targets in medicinal chemistry. These enzymes play a role in a number of human diseases, including type II diabetes and infection by Yersinia pestis, the causative agent of bubonic plague. Derivatives of squaric acids such as 2-aryl-1-hydroxycyclobut-1-ene-3,4-diones represent a new class of monoanionic inhibitors for PTPases.
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobutene-1,2-dione, ≥99.0% (HPLC)