MilliporeSigma
  • Pharmacological Characterization of the αvβ6 Integrin Binding and Internalization Kinetics of the Foot-and-Mouth Disease Virus Derived Peptide A20FMDV2.
Pharmacological Characterization of the αvβ6 Integrin Binding and Internalization Kinetics of the Foot-and-Mouth Disease Virus Derived Peptide A20FMDV2.
Pharmacology (2016-01-07)
Robert J Slack, Maryam Hafeji, Rebecca Rogers, Steve B Ludbrook, John F Marshall, David J Flint, Susan Pyne, Jane C Denyer
ABSTRACT
A20FMDV2 is a peptide derived from the foot-and-mouth disease virus with a high affinity and selectivity for the alpha-v beta-6 (αvβ6) arginyl-glycinyl-aspartic acid (RGD)-binding integrin. It has been shown to be an informative tool ligand in pre-clinical imaging studies for selective labelling of the αvβ6 integrin in a number of disease models. In a radioligand binding assay using a radiolabelled form of the peptide ([3H]A20FMDV2), its high affinity (K(D): 0.22 nmol/l) and selectivity (at least 85-fold) for αvβ6 over the other members of the RGD integrin family was confirmed. [3H]A20FMDV2 αvβ6 binding could be fully reversed only in the presence of EDTA, whereas a partial reversal was observed in the presence of excess concentrations of an RGD-mimetic small molecule (SC-68448) or unlabelled A20FMDV2. Using flow cytometry on bronchial epithelial cells, the ligand-induced internalization of αvβ6 by A20FMDV2 and latency-associated peptide-1 was shown to be fast (t(1/2): 1.5 and 3.1 min, respectively), concentration-dependent (EC50: values 1.1 and 3.6 nmol/l, respectively) and was followed by a moderately slow return of integrin to the surface. The results of the radioligand binding studies suggest that the binding of A20FMDV2 to the RGD-binding site on αvβ6 is required to maintain its engagement with the hypothesised A20FMDV2 synergy site on the integrin. In addition, there is evidence from flow cytometric studies that the RGD-ligand engagement of αvβ6 post-internalization plays a role in delaying recycling of the integrin to the cell surface. This mechanism may act as a homeostatic control of membrane αvβ6 following RGD ligand engagement.
Sigma-Aldrich
MISSION® esiRNA, targeting human ITGB6 (2)
Pricing and availability is not currently available.