MilliporeSigma
  • CA-125, but not galectin-3, complements CA 19-9 for discriminating ductal adenocarcinoma versus non-malignant pancreatic diseases.
CA-125, but not galectin-3, complements CA 19-9 for discriminating ductal adenocarcinoma versus non-malignant pancreatic diseases.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.] (2015-11-29)
Lucie Coppin, Kanza Benomar, François Corfiotti, Stéphane Cattan, Florence Renaud, Camille Lapere, Emmanuelle Leteurtre, Marie-Christine Vantyghem, Stéphanie Truant, Pascal Pigny
ABSTRACT
CA 19-9 is the gold standard biomarker of pancreatic adenocarcinoma despite several weaknesses in particular a high rate of false positives or negatives. CA-125 corresponding to MUC16 and galectin-3, a lectin able to interact with mucin-associated carbohydrates, are tumor-associated proteins. We investigated whether combined measurement of CA 19-9, galectin-3 and CA-125 may help to better discriminate pancreatic adenocarcinoma versus non-malignant pancreatic diseases. We evaluated by immunohistochemistry the expression of MUC4, MUC16 (CA-125) and galectin-3 in 31 pancreatic adenocarcinomas. We measured CA 19-9, CA-125 and Gal-3 in the serum from patients with pancreatic benign diseases (n = 58) or adenocarcinoma (n = 44). Clinical performance of the 3 biomarkers for cancer diagnosis and prognosis was analyzed. By immunohistochemistry, MUC16 and Gal-3 were expressed in 74% and 84% of adenocarcinomas versus 0% and 3.2% in peri-tumoral regions, respectively. At the serum level, CA 19-9 and CA125 were significantly higher in patients with pancreatic adenocarcinoma whereas Gal-3 levels did not differ. The performance of CA 19-9 for cancer detection was higher than those of CA-125 or Gal-3 by ROC analysis. However, CA-125 offers the highest specificity for malignancy (81%) because of an absence of false positives among type 2 diabetic patients. Cancer deaths assessed 6 or 12 months after diagnosis varied according to the initial CA-125 level (p < 0.006). Gal-3 is not an interesting biomarker for pancreatic adenocarcinoma detection. CA 19-9 alone exhibits the best performance but measuring CA-125 provides complementary information in terms of diagnosis and prognosis.